ENA300790662 Enamine

2-methyl-5H,6H,7H,8H-[1,2,4]triazolo[1,5-a]pyridin-6-amine

  • MDL number MFCD16817413
  • Cas Number : 1251925-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-63
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65540
Availability details for
Quantity
  • Availability
  • Add to Cart