ENA300790667 Enamine

1-(4-methyl-3-nitrophenyl)pyrrolidine-2,5-dione

  • MDL number MFCD01028285
  • Cas Number : 857753-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65632

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65632
Availability details for
Quantity
  • Availability
  • Add to Cart