ENA300790672 Enamine

2-(chloromethyl)-5-fluoro-1-(2,2,2-trifluoroethyl)-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD16817422
  • Cas Number : 1251924-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65774
Availability details for
Quantity
  • Availability
  • Add to Cart