ENA300790673 Enamine

N-(2-aminoethyl)-4-methyl-1,3-thiazole-5-carboxamide hydrochloride

  • MDL number MFCD16817423
  • Cas Number : 1251925-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65777
Availability details for
Quantity
  • Availability
  • Add to Cart