ENA300790674 Enamine

4-(3,4-difluorophenyl)butan-2-one

  • MDL number MFCD16990621
  • Cas Number : 1251925-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65783
Availability details for
Quantity
  • Availability
  • Add to Cart