ENA300790675 Enamine

2-fluoro-5-methyl-N-[1-(4-methylphenyl)ethyl]aniline hydrochloride

  • MDL number MFCD16817424
  • Cas Number : 1251922-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-65786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65786
Availability details for
Quantity
  • Availability
  • Add to Cart