ENA300790676 Enamine

2,6-bis(pyrrolidin-1-yl)aniline

  • MDL number MFCD16817425
  • Cas Number : 1172626-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65790

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65790
Availability details for
Quantity
  • Availability
  • Add to Cart