ENA300790679 Enamine

5-[(dimethylamino)methyl]-2-methylfuran-3-carboxylic acid hydrochloride

  • MDL number MFCD16817428
  • Cas Number : 1251922-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65821

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65821
Availability details for
Quantity
  • Availability
  • Add to Cart