ENA300790680 Enamine

1,2,3,4-tetrahydroquinoline-3-carboxamide

  • MDL number MFCD16817429
  • Cas Number : 4138-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65836
Availability details for
Quantity
  • Availability
  • Add to Cart