ENA300790681 Enamine

methyl 2-[(1,2,3,4-tetrahydroquinolin-8-yloxy)methyl]furan-3-carboxylate hydrochloride

  • MDL number MFCD16817430
  • Cas Number : 1251924-80-7
  • Molecular Weight : 323.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65875
Availability details for
Quantity
  • Availability
  • Add to Cart