ENA300790685 Enamine

2-chloro-N-methyl-N-[(1-methyl-1H-pyrazol-4-yl)methyl]acetamide hydrochloride

  • MDL number MFCD16817433
  • Cas Number : 1251925-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65978
Availability details for
Quantity
  • Availability
  • Add to Cart