ENA300790691 Enamine

2-[4-(1-aminoethyl)phenoxy]acetonitrile hydrochloride

  • MDL number MFCD16817441
  • Cas Number : 1251924-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66037
Availability details for
Quantity
  • Availability
  • Add to Cart