ENA300790693 Enamine

1,3-benzoxazole-5-carbonitrile

  • MDL number MFCD16817443
  • Cas Number : 132227-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66043
Availability details for
Quantity
  • Availability
  • Add to Cart