ENA300790694 Enamine

3-(chloromethyl)-5-cyclopropyl-4-methyl-4H-1,2,4-triazole hydrochloride

  • MDL number MFCD16817444
  • Cas Number : 1251923-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66050
Availability details for
Quantity
  • Availability
  • Add to Cart