ENA300790698 Enamine

4-{[(propan-2-yl)amino]methyl}benzonitrile hydrochloride

  • MDL number MFCD16817446
  • Cas Number : 1251923-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66084
Availability details for
Quantity
  • Availability
  • Add to Cart