ENA300790706 Enamine

N-(3,5-difluorophenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD16817454
  • Cas Number : 1251922-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66161
Availability details for
Quantity
  • Availability
  • Add to Cart