ENA300790715 Enamine

N-ethyl-2-methylcyclohexan-1-amine hydrochloride

  • MDL number MFCD11615758
  • Cas Number : 1251923-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233...235
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66303

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66303
Availability details for
Quantity
  • Availability
  • Add to Cart