ENA300790717 Enamine

2-(4-methylpiperazine-1-sulfonyl)acetic acid hydrochloride

  • MDL number MFCD16817465
  • Cas Number : 1251923-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66333
Availability details for
Quantity
  • Availability
  • Add to Cart