ENA300790718 Enamine

3-amino-N-(2-chloro-4-methylphenyl)propanamide hydrochloride

  • MDL number MFCD16817466
  • Cas Number : 1251924-84-1
  • Molecular Weight : 249.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66341
Availability details for
Quantity
  • Availability
  • Add to Cart