ENA300790719 Enamine

3-(3-aminophenyl)-1-phenylpropan-1-ol

  • MDL number MFCD16817467
  • Cas Number : 87082-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66347
Availability details for
Quantity
  • Availability
  • Add to Cart