ENA300790721 Enamine

3-amino-1-methyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD16817469
  • Cas Number : 54235-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66359
Availability details for
Quantity
  • Availability
  • Add to Cart