ENA300790727 Enamine

methyl N-(cyanomethyl)-N-cyclopropylcarbamate

  • MDL number MFCD16817477
  • Cas Number : 1251922-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66398
Availability details for
Quantity
  • Availability
  • Add to Cart