ENA300790728 Enamine

methyl 2-amino-2-[2-(trifluoromethyl)phenyl]acetate hydrochloride

  • MDL number MFCD16817478
  • Cas Number : 959139-69-6
  • Molecular Weight : 269.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66401
Availability details for
Quantity
  • Availability
  • Add to Cart