ENA300790729 Enamine

benzyl 3-(aminomethyl)benzoate hydrochloride

  • MDL number MFCD16817479
  • Cas Number : 1251924-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66402

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66402
Availability details for
Quantity
  • Availability
  • Add to Cart