ENA300790730 Enamine

phenyl N-(piperidin-4-yl)carbamate hydrochloride

  • MDL number MFCD16817480
  • Cas Number : 1251924-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66417
Availability details for
Quantity
  • Availability
  • Add to Cart