ENA300790731 Enamine

1-(5-amino-4H-1,2,4-triazol-3-yl)piperidine-4-carboxylic acid

  • MDL number MFCD16817481
  • Cas Number : 1251924-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66440
Availability details for
Quantity
  • Availability
  • Add to Cart