ENA300790732 Enamine

1-benzyl-3-hydrazinylpiperidine dihydrochloride

  • MDL number MFCD16817484
  • Cas Number : 1251924-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66473
Availability details for
Quantity
  • Availability
  • Add to Cart