ENA300790734 Enamine

2-amino-4-(methylsulfanyl)-N-(2-oxo-2,3-dihydro-1,3-benzoxazol-5-yl)butanamide hydrochloride

  • MDL number MFCD16817486
  • Cas Number : 1251922-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66517
Availability details for
Quantity
  • Availability
  • Add to Cart