ENA300790736 Enamine

2-oxo-1,2-dihydroquinoxaline-6-sulfonamide

  • MDL number MFCD16817488
  • Cas Number : 1251925-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66520
Availability details for
Quantity
  • Availability
  • Add to Cart