ENA300790738 Enamine

1-(6-chloropyridin-3-yl)-3-methylthiourea

  • MDL number MFCD16817489
  • Cas Number : 131748-90-8
  • Molecular Weight : 201.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66527
Availability details for
Quantity
  • Availability
  • Add to Cart