ENA300790739 Enamine

N-(3,4-dimethylphenyl)-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD16817490
  • Cas Number : 1251923-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66530
Availability details for
Quantity
  • Availability
  • Add to Cart