ENA300790742 Enamine

4-(4-ethyl-4H-1,2,4-triazol-3-yl)piperidine dihydrochloride

  • MDL number MFCD16817493
  • Cas Number : 1251923-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66547
Availability details for
Quantity
  • Availability
  • Add to Cart