ENA300790743 Enamine

N-(4-methyl-1,3-thiazol-2-yl)piperidin-4-amine dihydrochloride

  • MDL number MFCD16817494
  • Cas Number : 1251923-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66548

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66548
Availability details for
Quantity
  • Availability
  • Add to Cart