ENA300790746 Enamine

tert-butyl 3-[(propan-2-yl)amino]propanoate hydrochloride

  • MDL number MFCD16817497
  • Cas Number : 1251922-80-1
  • Molecular Weight : 223.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66566
Availability details for
Quantity
  • Availability
  • Add to Cart