ENA300790747 Enamine

(4-methoxy-3-nitrophenyl)methanesulfonyl fluoride

  • MDL number MFCD16817499
  • Cas Number : 1251924-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66583
Availability details for
Quantity
  • Availability
  • Add to Cart