ENA300790758 Enamine

3-amino-1-(3-ethynylphenyl)thiourea

  • MDL number MFCD16817510
  • Cas Number : 1071638-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66632
Availability details for
Quantity
  • Availability
  • Add to Cart