ENA300790759 Enamine

5-[(2-methoxyphenyl)methyl]-2,3-dihydro-1,3,4-oxadiazole-2-thione

  • MDL number MFCD11164449
  • Cas Number : 1094366-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66636

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66636
Availability details for
Quantity
  • Availability
  • Add to Cart