ENA300790767 Enamine

1-acetamido-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid

  • MDL number MFCD16817518
  • Cas Number : 1251924-59-0
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66728
Availability details for
Quantity
  • Availability
  • Add to Cart