ENA300790770 Enamine

2-(5,6-difluoro-1H-1,3-benzodiazol-1-yl)-3-methylbutanoic acid hydrochloride

  • MDL number MFCD16817521
  • Cas Number : 1251922-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66742
Availability details for
Quantity
  • Availability
  • Add to Cart