ENA300790771 Enamine

3-{4-methyl-3,6-dioxo-1H,2H,3H,6H,7H-pyrazolo[3,4-b]pyridin-5-yl}propanoic acid hydrochloride

  • MDL number MFCD16817522
  • Cas Number : 1251922-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66745
Availability details for
Quantity
  • Availability
  • Add to Cart