ENA300790772 Enamine

N1-(6-methylpyridazin-3-yl)ethane-1,2-diamine dihydrochloride

  • MDL number MFCD16817523
  • Cas Number : 1251925-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66751
Availability details for
Quantity
  • Availability
  • Add to Cart