ENA300790776 Enamine

2-(chloromethyl)-1-(2,2,2-trifluoroethyl)-1H-imidazole hydrochloride

  • MDL number MFCD16817527
  • Cas Number : 135206-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66765
Availability details for
Quantity
  • Availability
  • Add to Cart