ENA300790779 Enamine

6-(chloromethyl)-3-phenyl-5,6-dihydro-4H-1,2,4-oxadiazine

  • MDL number MFCD16817530
  • Cas Number : 56493-89-1
  • Molecular Weight : 210.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66784
Availability details for
Quantity
  • Availability
  • Add to Cart