ENA300790782 Enamine

5-(piperidin-4-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD16817533
  • Cas Number : 1046203-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66789
Availability details for
Quantity
  • Availability
  • Add to Cart