ENA300790783 Enamine

3-{[2-(4-methanesulfonylphenyl)-2-oxoethyl]carbamoyl}propanoic acid

  • MDL number MFCD16817534
  • Cas Number : 1251925-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66790
Availability details for
Quantity
  • Availability
  • Add to Cart