ENA300790787 Enamine

5-(2-cyclohexylethyl)-2,3-dihydro-1,3,4-oxadiazole-2-thione

  • MDL number MFCD10692292
  • Cas Number : 1152542-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66801
Availability details for
Quantity
  • Availability
  • Add to Cart