ENA300790799 Enamine

1-(3-chloro-4-fluorobenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD16817551
  • Cas Number : 1251923-60-0
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66892
Availability details for
Quantity
  • Availability
  • Add to Cart