ENA300790805 Enamine

2-({1,4,6-trimethyl-1H-pyrazolo[3,4-b]pyridin-3-yl}oxy)acetic acid

  • MDL number MFCD16817556
  • Cas Number : 1251924-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66920
Availability details for
Quantity
  • Availability
  • Add to Cart