ENA300790806 Enamine

1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one hydrochloride

  • MDL number MFCD16817557
  • Cas Number : 1251925-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66922

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66922
Availability details for
Quantity
  • Availability
  • Add to Cart