ENA300790808 Enamine

4-(3-chloropropyl)-1-methyl-1H-pyrazole hydrochloride

  • MDL number MFCD16817561
  • Cas Number : 1251925-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66932
Availability details for
Quantity
  • Availability
  • Add to Cart